missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Cup Liquid Sampler

Constructed of stainless steel and for sampling liquids and suspensions. Sampling Systems™ Cup Liquid Sampler enables discrete samples to be easily taken from a height within the product.

Brand:  Sampling Systems™ 5030J-1500

 View more versions of this product

Product Code. 11945437

  • 675.00 € / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 316L Stainless Steel Construction
  • Single point liquid sampler
Specifications

Specifications

Liquid Sampler
500 mL
1500 mm
Stainless Steel
80 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers