missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Cup Sampler

Manufactured from 316 stainless steel. The Cup Sampler enables discrete samples to be easily taken from known heights within the product.

Brand:  Sampling Systems™ 5030A-1500

Product Code. 11940018

  • 346.00 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Key Features:

  • Single point liquid sampler
  • Simple and easy operation
  • All product contact parts are made from 316L Stainless Steel
  • Variety of lengths and cup sizes available

Operation:

  • Insert the sampler to the required depth.
  • Squeeze handle which pushes the cup away from the lid, allowing the liquid to enter the cup.
  • Once the sample has been taken release the handle and remove from the product.
  • To empty the sampler either tip the sampler whilst squeezing the handle or unscrew the collecting cup.
Specifications

Specifications

Cup Sampler
100 mL
55 mm
Stainless Steel (316)
Reusable
1500 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.