Learn More
Applied Biosystems™ Custom 5' Fluorescent Labeled/Unlabeled Pairs
Custom 5' Fluorescent Labeled/Unlabeled Di-repeat + Tail
Brand:  Applied Biosystems™ 4304982
 View more versions of this product
Product Code. 15577280
  • 568.00 € / 300000pmol
Explore more special offers
Description
Description
A 5' fluorescent dye-labeled oligo and unlabeled complement are supplied in separate tubes. Choose among 6-FAM™, TET™, VIC™, HEX™, NED™, and PET™ as labels. Ideal for use in fragment analysis, gene expression studies, genotyping, and PCR. Oligos are desalted.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Custom 5' Labeled/Unlabeled Di-Repeat + Tail, 300000 pmol, 6-FAM™, TET™, VIC™, HEX™, NED™, PET™ 5' Reporter Dye, Choice of Delivered Scales, These Products can be used in a Variety of Applications
-20°C
Fragment Analysis, Gene Expression Studies, Genotyping, PCR
Custom 5' Labeled or Unlabeled Di-Repeat + Tail
300,000 pmol
Delivered either in Solution or Dried down