missing translation for 'onlineSavingsMsg'
Learn More

Value DNA Oligo , Desalted, Liquid, < 24 , 50 nmol

Custom DNA Oligo, 50N, Desalted, Liquid

Brand:  Invitrogen A15609

8.70 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 15450704

  • 13.38 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Value Oligos are the most cost-effective and fastest way to order oligonucleotides from us. Simply enter your sequence and then check out. Value Oligos are available at 25 nmol or 50 nmol scales, with a range of purification options to suit your needs.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DNA Oligo, 50 nmole, 50N, Liquid, Desalted, Cost-effective and Fastest
Desalted
Liquid
DNA Oligo
50 nmol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers