Learn More
Ambion™ Custom mirVana™ miRNA Inhibitor
Custom mirVana™ miRNA Inhibitor
Brand: Ambion™ 4464089
2603.46 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically inhibit endogenous miRNAs for the purpose of functional analysis via down-regulation. Use our GeneAssist™ miRNA Workflow Builder to design mimics and inhibitors for any miRNA sequence.Specifications
Room temperature | |
mirVana Custom miRNA Inhibitor, 250nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation | |
mirVana Custom miRNA Inhibitor | |
250 nmol |
For Research Use Only. Not for use in diagnostic procedures.