missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Custom mirVana™ miRNA Mimic

Custom mirVana™ miRNA Mimic

Brand:  Ambion™ 4464068

541.36 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15531124

  • 808.00 € / 5nmol

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

miRNA mimics are small, chemically modified double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity. Use our GeneAssist™ miRNA Workflow Builder to design both mimics and inhibitors for any miRNA sequence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer (-5 to -30°C).
miRNA
5 nmol
mirVana Custom miRNA Mimic, 5nmol, Lowest Off-Target Effects and Highest Potency, miRNA mimics are small, Chemically Modified Double-Stranded RNAs that mimic Endogenous miRNAs and Enable miRNA Functional Analysis by Up-Regulation of miRNA Activity
mirVana Custom miRNA Mimic
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.