Learn More
Ambion™ Custom RNA, Stealth, HPLC
Custom RNA, Stealth, HPLC
Brand:  Ambion™ 100001863
 View more versions of this product
Product Code. 10174283
  • 225.00 € / Each
Explore more special offers
Description
Description
Use this tool for fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks, and ready-to-clone BLOCK-iT™ miR RNAi Select.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Room temperature
RNA HPLC In-vivo
5 nmol
RNA HPLC In-vivo, 5nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation
HPLC