missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Custom Silencer™ siRNA

Product Code. p-7075785
Click to view available options
Quantity:
1 umol
100 nmol
20 nmol
40 nmol
Unit Size:
100nmol
1µmol
20nmol
40nmol
This item is not returnable. View return policy

Product Code. 15541144

Brand: Ambion™ AM16104

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Custom Silencer™ siRNA

RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at or below -20°C. Do not store in a frost-free freezer. (Dried oligonucleotides are shipped at ambient temperature.) Additional Materials Included: 1.75 mL Nuclease-free Water
Description Custom siRNA, 20nmol, Liquid, Standard Purity, Water Soluble, RNAi is the best Way to Effectively Knock Down Gene Expression to Study Protein Function in a Wide Range of Cell Types, Use our GeneAssist Custom siRNA Builder to Design and Order a Custom siRNA
RNAi Type siRNA
Sample Type RNA
Product Line Ambion™, Silencer™ Select
Product Type Custom siRNA
Purity Standard
Quantity 20 nmol
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado