Learn More
Applied Biosystems™ Custom TaqMan™ SNP Genotyping Assay, human
GREENER_CHOICE
Provide a highly flexible technology for detection of polymorphisms within any genome
Brand:  Applied Biosystems™ 4332072
488.58 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
 View more versions of this product
Product Code. 15511034
  • 958.00 € / Pack of 40
Explore more special offers
Includes: Two unlabelled PCR primers: forward and reverse (primers at 900nM final concentration); 1 VIC dye: MGB-labeled probe detects the Allele 1 sequence (probes at 200nM final concentration); 1 6FAM dye: MGB labeled probe detects the Allele 2 sequence (probes at 200nM final concentration)
Description
Description
Create assays by submitting confidential target sequences to Applied Biosystem's secure assay design pipeline using the Custom TaqMan Assay Design Tool. This pipeline has generated millions of assay designs by utilizing heuristic rules deduced from both manufacturing and assay performance data. These assays use TaqMan minor groove-binding (MGB) probes for superior allelic discrimination, improved signal-to-noise ratios, and design flexibility. All human SNP genotyping assays are functionally tested on 20 unique DNA samples to ensure assay amplification and sample clustering. These custom assays are designed, synthesized, formulated, optimized, and quality control tested. They are ideal for genotyping applications including screening, association, candidate region, candidate gene, or fine-mapping studies.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
40X
7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex ProFlex PCR System*, VeritiPro*, SimpliAmp*, MiniAmp*, Automated Thermal Cycler*
* If a thermal cycler is used for PCR amplification, the optional pre-read and the post-read must be performed separately on a real-time PCR
system in order to detect and record fluorescent signals.
5000 (384-well); 1000 (96-well)
Custom TaqMan Genotyping
Room Temperature
1 tube containing a 40X (S and M sizes) or 80X (L size) mix of pre-formulated assay (2 probes and 2 primers).
Store at -15 to -25°C.
Sustainable packaging
TaqMan™
M (1000 reactions), made to order