Learn More
Epredia™ Cut Microscope Slide
Premium slides manufactured through a unique proprietary cleaning process intended to provide consistent results and ease interpretation. This process allows these slides to be used in routine histology and microbiology applications.
Brand:  Epredia™ AA00000102E01MNZ20
Product Code. 17224884
  • 4.15 € / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Manufactured from Swiss extra-white soda-lime glass
  • Reduced iron content for low self-fluorescence
  • Greenish shimmer minimized
  • Guaranteed optimum, faultless transparency
  • Clean, flat and suitable for immediate use
Specifications
Specifications
Soda Lime Glass
ISO8037/I
No
1 mm
Uncoated
76 x 26 mm