missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cut Out Rings

With cut out section to allow easy removal of equipment.

24.95 €

Specifications

Material Mild Steel
Product Type Stativring
Description Cut out section
Finish Zinc Plated
View More Specs

Products 2
Product Code Brand Diameter (Metric) Inner Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Price Quantity & Availability  
10173744
View Documents
Fisherbrand™
2014/14
125 mm
24.95 €
Each
Estimated Shipment: 26-03-2025
to see stock.
Add to basket
 
10778675
View Documents
Fisherbrand™
2014/11
100 mm
24.95 €
Each
Estimated Shipment: 26-03-2025
to see stock.
Add to basket
 
Description

Description

  • Stem length to the center of bosshead is approximately 60mm
  • Constructed in mild steel with welded joints
  • Zinc-plated

Specifications

Specifications

Mild Steel
Stativring
Cut out section
Zinc Plated
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.