Learn More
QIAGEN Cutting Mat
For precise sample disc removal from FTA cards.
Brand: QIAGEN WB100020
This item is not returnable.
View return policy
Description
- The UniCore Cutting Mat ensures clean sample cuts from FTA/FTA Elute Cards and extends the life of the cutting tip of punch.
- The matt is are designed for repeated, extended use and can be cleaned with alcohol and is available in two formats.
Specifications
Green | |
Clean sample cuts from FTA/FTA™ Elute cards | |
1 Pk. |
Micro Punch | |
Cuttting Mat | |
Large (15.2 x 20.3 cm) |