missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Spare Cutting Wheel

Aids in cutting glass tubes.

Brand:  Fisherbrand™ 3406/1

Product Code. 13055983

  • 27.05 € / Pack of 5
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Manufactured in nickel plated zinc alloy
  • Accepts tubing up to 30mm in diameter
  • Replacement wheel is 10mm in diameter

Specifications

Specifications

Cutting Wheel Spare
Glass tubing cutter
10 mm
5 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.