Learn More
BTX™ Cuvette Rack
Cuvette Rack is an accesories for cuvettes.
Brand:  BTX™ 450208
Product Code. 15497290
  • 111.00 € / Each
Explore more special offers
Description
Description
  • 20 numbered positions
Specifications
Specifications
Storage and organisation of cuvettes while in use
Cuvette Rack
Rack Only (20 numbered positions)
Non-sterile
All standard format electroporation cuvettes
Cuvette Rack, 20 numbered positions
1 Ea.