missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CX 614

AMPAkine; positive allosteric modulator of AMPA

226.00 € - 915.00 €

Specifications

Purity >99%
Product Type CX 614
Molecular Weight (g/mol) 247.25
Inhibitors CX 614
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18765329
View Documents
Tocris Bioscience™
5149/10
10 mg
226.00 €
10mg
Please to purchase this item. Need a web account? Register with us today!
18716939
View Documents
Tocris Bioscience™
5149/50
50 mg
915.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AMPAkine; positive allosteric modulator of AMPA. Prototypical benzamide which occupies subsite A only. Modulates channel deactivation and desensitization.
Specifications

Specifications

>99%
CX 614
247.25
CX 614
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only