missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Cy™3B Mono-reactive Maleimide Fluorescent Dyes

Product Code. 11525834 Shop All Cytiva Products
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
missing translation for 'unitSize'
1mg
This item is not returnable. View return policy

Product Code. 11525834

missing translation for 'mfr': Cytiva PA63131

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Combines exceptional photostability with bright and intense signals. Cytiva Cy3B Maleimides Mono-Reactive CyDye is a more selective method of antibody labeling that reduces the possibility of the label interfering with subsequent antibody-antigen reactions.

CyDye™ fluors are versatile fluorophores designed for a broad range of applications. CyDye™ fluors produce high sensitivity and highly reproducible performance at a biologically suitable pH range of 3 to 10.

Cy™3B Maleimides is a more selective method of antibody labeling that reduces the possibility of the label interfering with subsequent antibody-antigen reactions.

  • The use of multicolor CyDye™ flours enables multiplexing without crosstalk for high accuracy
  • CyDye™ flours readily dissolve in aqueous buffers, which eliminates the need of organic solvents
  • High purity and thorough QC testing generate high levels of chromophore and reactive dye content
  • Narrow excitation and emission bands result in discrete signals from each fluor
TRUSTED_SUSTAINABILITY

Specifications

Formulation 100% Cy3B (maleimide)
Format Solid
Quantity 1 mg
Product Type Cy3B Maleimide
Molecular Weight (g/mol) 796
Solubility Soluble in cold and hot water
Label or Dye Cy3B
Color Red
Emission 570
Excitation 559
compliance-icons compliance-icons
Signal Word
  • Danger||
Hazard Category
  • Acute toxicity Category 3
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.