Learn More
Cytiva Cy7 Bis NHS Ester
Suitable for general labeling of amine residues
Brand:  Cytiva PA17002
Product Code. 10440315
  • 9800.00 € / 50mg
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The packs contain fluors carrying two reactive groups on each dye molecule for general labeling of protein amine groups.
  • Suitable for general labeling of amine residues.
  • Large range of bulk packs available for greatest economy.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100% Cy7 (Bis NHS ester)
50 mg
2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
Purple
Used for high-throughput screening (HTS) applications
Cy7 Bis NHS Ester
Cy7
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 2