missing translation for 'onlineSavingsMsg'
Learn More

Cyanamide, 95%, stabilized, Thermo Scientific Chemicals

56.60 € - 518.00 €

Chemical Identifiers

CAS 420-04-2
Molecular Formula CH2N2
Molecular Weight (g/mol) 42.04
MDL Number MFCD00007572
InChI Key XZMCDFZZKTWFGF-UHFFFAOYSA-N
Synonym hydrogen, carbimide, amidocyanogen, carbamonitrile, cyanoamine, cyanogenamide, cyanogen nitride, n-cyanoamine, alzogur, dormex
PubChem CID 9864
ChEBI CHEBI:16698
IUPAC Name cyanamide
SMILES C(#N)N
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10130412
View Documents
Thermo Scientific Acros
181950250
25 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10110462
View Documents
Thermo Scientific Acros
181951000
100 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10135823
View Documents
Thermo Scientific Acros
181955000
500 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

420-04-2
42.04
XZMCDFZZKTWFGF-UHFFFAOYSA-N
9864
cyanamide
CH2N2
MFCD00007572
hydrogen, carbimide, amidocyanogen, carbamonitrile, cyanoamine, cyanogenamide, cyanogen nitride, n-cyanoamine, alzogur, dormex
CHEBI:16698
C(#N)N

Specifications

420-04-2
White
141°C
94% min. (Argentometry)
H2NCN
03, IV, 145
15, 2674
Solubility in water: 775g/L. Other solubilities: soluble in alchol,phenols,amines,ethers,ketones,slightly soluble in benzene and halogenated,hydrocarbons
C(#N)N
0.1% (typically) formic acid
9864
42.04
Crystals
40.0°C to 47.0°C
260.0°C
Authentic
CH2N2
MFCD00007572
01,170; 03,71; 05,168; 12,147
hydrogen, carbimide, amidocyanogen, carbamonitrile, cyanoamine, cyanogenamide, cyanogen nitride, n-cyanoamine, alzogur, dormex
XZMCDFZZKTWFGF-UHFFFAOYSA-N
cyanamide
42.04
CHEBI:16698
95%
Cyanamide, 95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic in contact with skin.
Suspecte

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 206-992-3

Recommended Storage : Refrigerator +4°C

Product Identifier
  • Cyanamide, stabilized

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P361-Take off immediately all contaminated clothing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.