Learn More
Cyanogen bromide, 97%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 506-68-3 |
---|---|
Molecular Formula | CBrN |
MDL Number | MFCD00011597 |
InChI Key | ATDGTVJJHBUTRL-UHFFFAOYSA-N |
Synonym | cyanogen bromide, bromine cyanide, bromocyanide, cyanobromide, bromocyanogen, bromocyan, campilit, cyanogen monobromide, bromure de cyanogen, cyanogen bromide brcn |
PubChem CID | 10476 |
IUPAC Name | carbononitridic bromide |
SMILES | C(#N)Br |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
506-68-3 | |
MFCD00011597 | |
cyanogen bromide, bromine cyanide, bromocyanide, cyanobromide, bromocyanogen, bromocyan, campilit, cyanogen monobromide, bromure de cyanogen, cyanogen bromide brcn | |
carbononitridic bromide |
CBrN | |
ATDGTVJJHBUTRL-UHFFFAOYSA-N | |
10476 | |
C(#N)Br |
Specifications
506-68-3 | |
1.44 | |
CBrN | |
MFCD00011597 | |
15, 2683 | |
ATDGTVJJHBUTRL-UHFFFAOYSA-N | |
carbononitridic bromide | |
10476 | |
≥96.0% | |
Cyanogen bromide, 97% |
White | |
97% | |
BrCN | |
01,174; 02,93; 04,110; 05,169; 06,148; 08,131; 09,127 | |
cyanogen bromide, bromine cyanide, bromocyanide, cyanobromide, bromocyanogen, bromocyan, campilit, cyanogen monobromide, bromure de cyanogen, cyanogen bromide brcn | |
C(#N)Br | |
105.922 | |
105.93 | |
Crystals |
Safety and Handling
GHS H Statement:
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
Recommended Storage : Refrigerator +4°C
- Cyanogen bromide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H310-Fatal in contact with skin.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H300-Fatal if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only