missing translation for 'onlineSavingsMsg'
Learn More

Cyanogen iodide, 98%, Thermo Scientific Chemicals

Product Code. 10006172 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10006172

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-78-5
Molecular Formula CIN
InChI Key WPBXOELOQKLBDF-UHFFFAOYSA-N
Synonym cyanogen iodide, iodine cyanide, jodcyan, iodocyanide, iodine monocyanide, cyanogen monoiodide, iodine cyanide i cn, unii-81esv254uf, cyanic iodide, cyano iodide
PubChem CID 10478
IUPAC Name carbononitridic iodide
SMILES C(#N)I

Specifications

Melting Point 142.0°C to 149.0°C
Color Beige to Brown or Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Glass bottle
Linear Formula ICN
Quantity 5 g
Merck Index 15,2685
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in diethyl ether,ethanol,ether,alcohols,and volatile oils
Molecular Weight (g/mol) 152.92
Formula Weight 152.92
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Cyanogen iodide
Show More Show Less
compliance-icons
Product Identifier
  • Cyanogen iodide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.