missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Non-Sterile CyChlor
BIOPHARMA

Provided ready-to-use and is efficacious against bacteria and yeasts in 3 mins. Contec™ Non-Sterile CyChlor is a new broad spectrum disinfectant designed for every day use.

Brand:  Contec™ FBC500CC

82.29 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 15939875

  • 118.00 € / 2 bottles
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Broad spectrum efficacy in 3 mins saves time spent on biodecontamination.
  • Filtered to 0.2 microns and filled in a Grade A environment ensuring the product is free from contamination and particulates
  • Contains no quaternary ammonium or surfactant so it's very low residue, saving time on residue removal
  • No hazard classification so only basic PPE required and no special disposal is necessary
  • Trigger spray can be set to jet or spray - large droplet size reduces the risk of inhalation and provides good surface coverage
  • Double bagged packed in linear tear packaging so each bag is easy to open even when wearing gloves
  • Not classed as corrosive and can be used safely in all areas of the cleanroom — this needs a bit of random text deleting from the end
Specifications

Specifications

Disinfectant
3 to 6
Non-sterile
Not Autoclavable
Fluid
Limited-use
No
5 L
Liquid
Unscented
Colorless
Clear
Capped Container
Filtered
2/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers