missing translation for 'onlineSavingsMsg'
Learn More

Cyclamic acid, MedChemExpress

MedChemExpress Cyclamic acid (Cyclohexylsulfamic acid) is one of the most widely used artificial sweetenersin food and pharmaceuticals.

Encompass_Preferred

POA - 112.05 €

Specifications

Chemical Name or Material Cyclamic acid
Physical Form Solid
CAS 100-88-9
Color White
Purity Grade Notes Research
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16492219
Encompass_Preferred
View Documents
Medchem Express
HY-B0541
1 g
112.05 €
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16420988
Encompass_Preferred
View Documents
Medchem Express
HY-B0541
5 g
POA
5g
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

Cyclamic acid (Cyclohexylsulfamic acid) is one of the most widely used artificial sweetenersin food and pharmaceuticals.
Specifications

Specifications

Cyclamic acid
100-88-9
Research
Cyclohexylsulfamic acid Cyclamate
O=S(O)(NC1CCCCC1)=O
179.24
Research
Solid
White
C6H13NO3S
DMSO : 100 mg/mL (557.91 mM; Need ultrasonic)
179.24
98.0%
Videos
Safety and Handling

Safety and Handling

H315∣H319

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Cyclamic acid, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.