missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Cyclin D1/Bcl-1, Rabbit Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Cyclin D1/Bcl-1, Rabbit Monoclonal Antibody.

224.00 € - 1080.00 €

Specifications

Antigen Cyclin D1/Bcl-1
Clone SP4
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Product Code Brand Form Quantity Price Quantity & Availability  
Product Code Brand Form Quantity Price Quantity & Availability  
10733436
View Documents
Epredia™
RM-9104-S0
Concentrated 100 μL
224.00 €
100µL
Please to purchase this item. Need a web account? Register with us today!
11594564
View Documents
Epredia™
RM-9104-S1
Concentrated 500 μL
533.00 €
100µL
Please to purchase this item. Need a web account? Register with us today!
12653687
View Documents
Epredia™
RM-9104-S
Concentrated 1 mL
1080.00 €
1mL
Please to purchase this item. Need a web account? Register with us today!
12643687
View Documents
Epredia™
RM-9104-RQ
Ready-to-Use 12 mL
708.00 €
12mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cyclin D1 or PRAD-1 or bcl-1 is one of the key cell cycle regulators, and functions in association with cdk4 and/or cdk6 by phosphorylating the Rb protein. It is a putative proto-oncogene overexpressed in a wide variety of human neoplasms including mantle cell lymphomas (MCL).

Host Species: Rabbit

Clone: SP4

Isotype: IgG

Species Reactivity: Human, Mouse, Rat. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide from C-terminus of human cyclin D1

Molecular Weight: 36 kDa

Positive Control: MAD109 cells, breast CA or mantle cell lymphoma

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Specifications

Specifications

Cyclin D1/Bcl-1
Western Blot
Unconjugated
IVD
A synthetic peptide from C-terminus of human cyclin D1
SP4
Monoclonal
Rabbit
Cell Cycle
Primary
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • APD Primary Antibody Products, Antibody Diluent

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P363-Wash contaminated clothing before reuse.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.