Learn More
Cyclobutylamine, 98%, Thermo Scientific Chemicals
59.10 € - 795.00 €
Chemical Identifiers
CAS | 2516-34-9 |
---|---|
Molecular Formula | C4H9N |
Molecular Weight (g/mol) | 71.123 |
MDL Number | MFCD00001328 |
InChI Key | KZZKOVLJUKWSKX-UHFFFAOYSA-N |
Synonym | cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride |
PubChem CID | 75645 |
IUPAC Name | cyclobutanamine |
SMILES | C1CC(C1)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11405463
|
Thermo Scientific Alfa Aesar
A13423.03 |
1 g |
59.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11415463
|
Thermo Scientific Alfa Aesar
A13423.06 |
5 g |
227.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11425463
|
Thermo Scientific Alfa Aesar
A13423.14 |
25 g |
795.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Substrate employed in a microwave-assisted synthesis of 7-azaindoles from dihalopyridines. Also it is a reactant in the preparation of imidazo[1,2-b]pyridazine derivatives as selective and orally available Mps1 (TTK) kinase inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSubstrate employed in a microwave-assisted synthesis of 7-azaindoles from dihalopyridines. Also it is a reactant in the preparation of imidazo[1,2-b]pyridazine derivatives as selective and orally available Mps1 (TTK) kinase inhibitors.
Solubility
Slightly soluble in water.
Notes
Hygroscopic, store way form water/moisture. Store in cool, dry place in tightly closed container. Store away from oxidizing agents.
Chemical Identifiers
2516-34-9 | |
71.123 | |
KZZKOVLJUKWSKX-UHFFFAOYSA-N | |
75645 | |
C1CC(C1)N |
C4H9N | |
MFCD00001328 | |
cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride | |
cyclobutanamine |
Specifications
2516-34-9 | |
82°C to 83°C | |
C4H9N | |
MFCD00001328 | |
2069297 | |
cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride | |
KZZKOVLJUKWSKX-UHFFFAOYSA-N | |
cyclobutanamine | |
75645 | |
98% |
0.836 | |
−4°C (24°F) | |
1.4376 | |
UN2733 | |
Air sensitive; Hygroscopic | |
Slightly soluble in water. | |
C1CC(C1)N | |
71.123 | |
71.12 | |
Cyclobutylamine |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 219-736-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only