missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclobutylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
This compound can be employed for cross coupling with acid chlorides, aryl halides, and substituted or non-substituted allyl halides. They may also undergo Michael Addition to unsaturated ketones or additions to alkynes to give subs
This compound can be employed for cross coupling with acid chlorides, aryl halides, and substituted or non-substituted allyl halides. They may also undergo Michael Addition to unsaturated ketones or additions to alkynes to give subs
Especificaciones
Especificaciones
| CAS | 1019205-65-2 |
| Density | 0.984 |
| Molecular Formula | C4H7BrZn |
| MDL Number | MFCD09801445 |
| Quantity | 50 mL |
| UN Number | UN3399 |
| Sensitivity | Air sensitive |
| Synonym | cyclobutylzinc bromide, cyclobutylzinc bromide, 0.5 m in thf, cyclobutylzinc bromide 0.5m in tetrahydrofuran, cyclobutylzinc bromide, 0.5m in tetrahydrofuran |
| Solubility Information | Reacts with water. |
| InChI Key | WNFQEBSVYUDIES-UHFFFAOYSA-M |
| Mostrar más |
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido