missing translation for 'onlineSavingsMsg'
Learn More

Cyclododecanone, 99+%, Thermo Scientific Chemicals

Product Code. 10500371
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 830-13-7
Molecular Formula C12H22O
Molecular Weight (g/mol) 182.31
MDL Number MFCD00003722
InChI Key SXVPOSFURRDKBO-UHFFFAOYSA-N
Synonym cyclododecan-1-one, dsstox_cid_7322, dsstox_rid_78407, dsstox_gsid_27322, cyclododecanon, cdon, cyclododecanone, acmc-209pr1, ksc448s4j
PubChem CID 13246
IUPAC Name cyclododecanone
SMILES C1CCCCCC(=O)CCCCC1

Specifications

Melting Point 59.0°C to 61.0°C
Color Colorless to White
Boiling Point 277.0°C
Flash Point 118°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C12H22(=O)
Quantity 100 g
Beilstein 07, II, 48
Solubility Information Solubility in water: insoluble.
Viscosity 4.28 mPa.s (70°C)
Formula Weight 182.31
Percent Purity 99+%
Physical Form Adhering Crystals
Chemical Name or Material Cyclododecanone, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Cyclododecanone
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'