Learn More
Cycloguanil-d6, MedChemExpress
MedChemExpress Cycloguanil D6 is the deuterium labeled Cycloguanil, which is a dihydrofolate reductase inhibitor.
Brand: Medchem Express HY-12784S
This item is not returnable.
View return policy
Description
Cycloguanil D6 is the deuterium labeled Cycloguanil, which is a dihydrofolate reductase inhibitor.Specifications
Cycloguanil-d6 | |
White | |
C11H8D6ClN5 | |
ClC1=CC=C(N2C(N)=NC(N)=NC2(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C1 | |
257.75 | |
Research |
1 mg | |
Research | |
Chlorguanide triazine-d6 | |
257.75 | |
98.0% | |
Solid |