missing translation for 'onlineSavingsMsg'
Learn More
Cycloheptyl bromide, 97%, Thermo Scientific Chemicals Product Code.: 10616682

Cycloheptyl bromide, 97%, Thermo Scientific Chemicals

Product Code. 10616682
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10616682

Brand: Thermo Scientific Acros 111081000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2404-35-5
Molecular Formula C7H13Br
Molecular Weight (g/mol) 177.09
MDL Number MFCD00004149
InChI Key LOXORFRCPXUORP-UHFFFAOYSA-N
Synonym cycloheptane, bromo, cycloheptyl bromide, cycloheptylbromide, bromocyclo-heptane, 2-bromocycloheptane, bromocycloheptane, acmc-209g8c
PubChem CID 16992
IUPAC Name bromocycloheptane
SMILES BrC1CCCCCC1

Specifications

Color Yellow
Density 1.2800g/mL
Boiling Point 71°C to 72°C (10.0 mmHg)
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C7H13Br
Refractive Index 1.5035 to 1.5055
Quantity 100 g
Beilstein 05, 29
Specific Gravity 1.28
Formula Weight 177.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cycloheptyl bromide, 97%
Show More Show Less
Product Identifier
  • Cycloheptyl bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cycloheptyl bromide, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.