missing translation for 'onlineSavingsMsg'
Learn More
Riedel-de Haën Cyclohexane Product Code.: 15975460
compliance-icons compliance-icons compliance-icons compliance-icons

Riedel-de Haën Cyclohexane

Product Code. 15975460
45 L
Click to view available options
Quantity:
18 L
45 L
200 L
This item is not returnable. View return policy

Product Code. 15975460

Brand: Riedel-de Haën 3311745LRC

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Cyclohexane, Puriss. p.a., ACS Reagent, ≥99.5% (GC).

  • ACS general use solvents.
  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis.
  • Suitable for less demanding applications such as routine isocratic HPLC.
  • Variations and decrease issues traceable to solvent contamination.
  • All solvents are tested to meet ACS general use specifications.

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym Hexahydrobenzene
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

pH Not applicable
Melting Point 6.4°C
Color Colorless
CAS Max % ≤100.00
Density 0.78 g/cm3 (20°C)
Boiling Point 81°C (1.013 hPa)
Quantity 45 L
Flash Point 8.3 %(V)
Packaging Returnable Stainless Steel Drum
Physical Form Liquid
Product Line Puriss. p.a.
Linear Formula C6H12
Vapor Pressure 130 hPa (25°C)
UN Number UN1145
Viscosity 0.94 mPa.s (20°C)
Beilstein 1900225
Concentration ≥99.50%
Percent Purity ≥99.50%
Grade ACS
Chemical Name or Material Cyclohexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Riedel-de Haën Cyclohexane >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.