missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, ≥99.8%, Laboratory Reagent, Honeywell™

Product Code. 15667720
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
25 L
4 x 2.5 L
4 x 5 L
150 kg
Packaging:
Aluminum Bottle
Glass Bottle
Metal Can
Metal Drum
Stainless Steel Can
This item is not returnable. View return policy
This item is not returnable. View return policy

Laboratory Reagent,≥99.8%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.4°C
Color Colorless
Density 0.779g/mL at 25°C
Boiling Point 81°C (1013 hPa)
Quantity 4 x 5 L
Explosion Limit 0.09
Ignition Point 500°F
Odor Characteristic
Packaging Aluminum Bottle
Physical Form Liquid
Vapor Pressure 168.8mmHg (37.7°C), 77mmHg (20°C)
UN Number UN1145
Beilstein 1900225
Formula Weight 84.16g/mol
Percent Purity ≥99.8%
Chemical Name or Material Cyclohexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'