missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, 99+%, Thermo Scientific Chemicals

Product Code. 11413087
Click to view available options
Quantity:
1000 mL
5000 mL

Cyclohexane is used to prepare cyclohexanone-cyclohexanol mixture (i.e. KA oil) by catalytic oxidation, which finds application as raw material for adipic acid and caprolactam. It is used as an organic solvent, paint remover and involved in the calibration of differential scanning calorimetry (DSC) instruments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexane is used to prepare cyclohexanone-cyclohexanol mixture (i.e. KA oil) by catalytic oxidation, which finds application as raw material for adipic acid and caprolactam. It is used as an organic solvent, paint remover and involved in the calibration of differential scanning calorimetry (DSC) instruments.

Solubility
Miscible with ether, alcohol, acetone and olive oil. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.5°C
Density 0.779
Boiling Point 80°C to 81°C
Quantity 5000 mL
Flash Point −18°C (−1°F)
Odor Pungent
Refractive Index 1.426
UN Number UN1145
Beilstein 1900225
Merck Index 14,2723
Solubility Information Miscible with ether,alcohol,acetone and olive oil. Immiscible with water.
Formula Weight 84.16
Percent Purity ≥99%
Chemical Name or Material Cyclohexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Consigli di prudenza
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.
Informazioni supplementari
  • MIXTURE LIST-Contains : Cyclohexane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'