missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane, Certified AR for Analysis, Fisher Chemical™
C6H12, CAS Number-110-82-7, 2.5L, 81 deg.C, CHEBI:29005, Colorless, 84.15g/mol, XDTMQSROBMDMFD-UHFFFAOYSA-N, cyclohexane, 6.5 deg.C, 3814, 84.162, Amber glass bottle, Liquid, 8078, C1CCCCC1, 2.9, 104mbar at 20 deg.C, 0.94 mPaS at 20 deg.C
Brand: Fisher Chemical C/8921/15
46.78 EUR valid until 2024-12-31
Use promo code "21665" to get your promotional price.
This item is not returnable.
View return policy
Description
C6H12, CAS Number-110-82-7, 2.5L, 81 deg.C, CHEBI:29005, Colorless, 84.15g/mol, XDTMQSROBMDMFD-UHFFFAOYSA-N, cyclohexane, 6.5 deg.C, 3814, 84.162, Amber glass bottle, Liquid, 8078, C1CCCCC1, 2.9, 104mbar at 20 deg.C, 0.94 mPaS at 20 deg.C
Specifications
6.5°C | |
Colorless | |
1 L | |
Liquid | |
MFCD00003814 | |
0.94 mPaS at 20°C | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
cyclohexane | |
8078 | |
84.15g/mol |
110-82-7 | |
81°C | |
Amber Glass Bottle | |
C6H12 | |
104mbar at 20°C | |
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
C1CCCCC1 | |
84.16 | |
CHEBI:29005 |