missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, Certified AR for Analysis, Fisher Chemical™

Product Code. 10724371 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic can
Plastic-coated Glass Bottle
Unit Size:
1 L
2.5 L
25 L
5 L
This item is not returnable. View return policy

Product Code. 10724371

Brand: Fisher Chemical C/8921/PB17

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C6H12, CAS Number-110-82-7, 2.5L, 81 deg.C, CHEBI:29005, Colorless, 84.15g/mol, XDTMQSROBMDMFD-UHFFFAOYSA-N, cyclohexane, 6.5 deg.C, 3814, 84.162, Amber glass bottle, Liquid, 8078, C1CCCCC1, 2.9, 104mbar at 20 deg.C, 0.94 mPaS at 20 deg.C

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 6.5°C
CAS 110-82-7
Color Colorless
Boiling Point 81°C
Quantity 2.5 L
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Molecular Formula C6H12
MDL Number 3814
Vapor Pressure 104 mbar at 20°C
Viscosity 0.94 mPaS at 20°C
Formula Weight 84.15g/mol
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.