missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane, CHROMASOLV™, for HPLC, ≥99.7%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
100 mL
1 L
2 L
2.5 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
Melting Point | 4°C to 7°C (literature) |
Absorbance | max. 0.10 at 235 nm, max. 0.20 at 230 nm, max. 0.50 at 220 nm, max. 1.00 at 210 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm, max. 0.01 at 255 nm, max. 0.03 at 250 nm, max. 0.08 at 240 nm |
CAS Max % | ≤100.0000% |
Linear Formula | C6H12 |
UN Number | UN1145 |
Beilstein | 1900225 |
Percent Purity | ≥99.7% |
Grade | HPLC |
Density | 0.779 g/mL at 25 °C(lit.); 0.780 g/mL at 25 °C |
Boiling Point | 81°C |
Show More |
Safety and Handling
Product Identifier
- Cyclohexane
- Danger
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'