missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, HPLC Grade, J.T. Baker™

Product Code. 15578564
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High purity solvents for use in analytical and preparative separations

    For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry

Specifications

CAS 110-82-7
Density 1.318g/mL at 25°C (typical)
Boiling Point 39.5°C (typical)
Quantity 4 L
Flash Point −18.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.78kg
Molecular Formula C6H12
MDL Number MFCD00003814
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
SMILES C1CCCCC1
IUPAC Name cyclohexane
Molecular Weight (g/mol) 84.16
PubChem CID 8078
ChEBI CHEBI:29005
Formula Weight 84.16
Grade HPLC
Chemical Name or Material Cyclohexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P331-Do NOT induce vomiting.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'