Learn More
Cyclohexanol, 98%, Thermo Scientific Chemicals
55.58 € - 319.59 €
Chemical Identifiers
CAS | 108-93-0 |
---|---|
Molecular Formula | C6H12O |
Molecular Weight (g/mol) | 100.16 |
InChI Key | HPXRVTGHNJAIIH-UHFFFAOYSA-N |
Synonym | cyclohexyl alcohol, hexahydrophenol, hydrophenol, hydroxycyclohexane, hexalin, 1-cyclohexanol, hydralin, adronal, naxol, adronol |
PubChem CID | 7966 |
ChEBI | CHEBI:18099 |
IUPAC Name | cyclohexanol |
SMILES | C1CCC(CC1)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10377120
|
Thermo Scientific Acros
147680010 |
1 L | Glass Bottle |
55.58 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10696342
|
Thermo Scientific Acros
147680025 |
2.5 L | Glass Bottle | N/A | |||||
11944641
|
Thermo Scientific Acros
147680100 |
10 L | Plastic drum |
319.59 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-93-0 | |
100.16 | |
cyclohexyl alcohol, hexahydrophenol, hydrophenol, hydroxycyclohexane, hexalin, 1-cyclohexanol, hydralin, adronal, naxol, adronol | |
CHEBI:18099 | |
C1CCC(CC1)O |
C6H12O | |
HPXRVTGHNJAIIH-UHFFFAOYSA-N | |
7966 | |
cyclohexanol |
Specifications
108-93-0 | |
Colorless | |
161.0°C | |
Authentic | |
C6H12O | |
1.4630 to 1.4670 | |
15,2718 | |
cyclohexyl alcohol, hexahydrophenol, hydrophenol, hydroxycyclohexane, hexalin, 1-cyclohexanol, hydralin, adronal, naxol, adronol | |
HPXRVTGHNJAIIH-UHFFFAOYSA-N | |
cyclohexanol | |
7966 | |
100.16 | |
Liquid After Melting |
23.0°C | |
0.9600g/mL | |
67°C | |
98% | |
C6H11OH | |
06,5 | |
0.96 | |
Solubility in water: 3.6g/100mL (20°C). Other solubilities: soluble in ethanol, ethyle acetate and, aromatic hydrocarbons | |
C1CCC(CC1)O | |
100.16 | |
CHEBI:18099 | |
98% | |
Cyclohexanol |
Safety and Handling
GHS H Statement
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protec
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-630-6
- Cyclohexanol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only