missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexene, 99%, pure, stabilized, Thermo Scientific Chemicals

Product Code. 10173200
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-83-8
InChI Key HGCIXCUEYOPUTN-UHFFFAOYSA-N
Synonym tetrahydrobenzene, cyclohex-1-ene, benzene tetrahydride, 1,2,3,4-tetrahydrobenzene, benzenetetrahydride, hexanaphthylene, 1-cyclohexene, cykloheksen, benzene, tetrahydro, 3,4,5,6-tetrahydrobenzene
PubChem CID 8079
ChEBI CHEBI:36404
IUPAC Name cyclohexene
SMILES C1CCC=CC1

Specifications

Melting Point -104.0°C
Color Colorless
Density 0.8100g/mL
Boiling Point 83.0°C
Flash Point −20°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H10
Linear Formula C6H10
Refractive Index 1.4450 to 1.4470
Quantity 500 mL
Beilstein 05,63
Merck Index 15,272
Specific Gravity 0.81
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, miscible with all common organic solvents
Molecular Weight (g/mol) 82.13
Viscosity 0.66 mPa.s (22°C)
Formula Weight 82.13
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material Cyclohexene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.