missing translation for 'onlineSavingsMsg'
Learn More
Cyclohexene oxide, 98%, Thermo Scientific Chemicals Product Code.: 10510371

Cyclohexene oxide, 98%, Thermo Scientific Chemicals

Product Code. 10510371
100 mL, Glass bottle
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10510371

Brand: Thermo Scientific Acros 111211000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 286-20-4
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.14
MDL Number MFCD00005162
InChI Key ZWAJLVLEBYIOTI-UHFFFAOYSA-N
Synonym cyclohexene oxide, 7-oxabicyclo 4.1.0 heptane, 1,2-epoxycyclohexane, cyclohexene epoxide, cyclohexylene oxide, tetramethyleneoxirane, cyclohexene 1-oxide, epoxycyclohexane, cyclohexeneoxide, 1,2-cyclohexene oxideShow MoreShow Less
PubChem CID 9246
IUPAC Name 7-oxabicyclo[4.1.0]heptane
SMILES C1CCC2C(C1)O2

Specifications

Melting Point -40.0°C
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 129.0°C to 130.0°C
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4520 to 1.4540
Quantity 100 mL
Beilstein 17, 21
Specific Gravity 0.97
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol, ether and acetone, soluble in most other organic solvents
Viscosity 1.75 mPa.s (25°C)
Formula Weight 98.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclohexene oxide, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0457806 Certificate TypeCertificate of Analysis Date16/11/2023 Catalog Number111211000,111215000
Lot NumberA0451551 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number111211000,111215000
Lot NumberA0405651 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number111211000,111215000
Lot NumberA0407792 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number111211000,111215000
Lot NumberA0437282 Certificate TypeCertificate of Analysis Date16/12/2021 Catalog Number111211000,111215000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexene oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclohexene oxide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.