missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexene oxide, 98+%, Thermo Scientific Chemicals

Product Code. 11474584
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, dyestuffs and in agrochemical industries. It is used in medicine. Cyclohexene oxide (epoxycyclohexane) is an useful monomer in polymerization and coating industry. It is used in the synthesis of alicyclic target materials including pharmaceuticals, perfumery and dyestuffs. It is used as a monomer in photopolymerizations, with carbonmonoxide to yield aromatic polycarbonates which has minimum impurities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, dyestuffs and in agrochemical industries. It is used in medicine. Cyclohexene oxide (epoxycyclohexane) is an useful monomer in polymerization and coating industry. It is used in the synthesis of alicyclic target materials including pharmaceuticals, perfumery and dyestuffs. It is used as a monomer in photopolymerizations, with carbonmonoxide to yield aromatic polycarbonates which has minimum impurities.

Solubility
Insoluble in water

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 286-20-4
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00005162
InChI Key ZWAJLVLEBYIOTI-UHFFFAOYSA-N
Synonym cyclohexene oxide, 7-oxabicyclo 4.1.0 heptane, 1,2-epoxycyclohexane, cyclohexene epoxide, cyclohexylene oxide, tetramethyleneoxirane, cyclohexene 1-oxide, epoxycyclohexane, cyclohexeneoxide, 1,2-cyclohexene oxide
PubChem CID 9246
IUPAC Name 7-oxabicyclo[4.1.0]heptane
SMILES C1CCC2C(C1)O2

Specifications

Melting Point -40°C
Density 0.972
Boiling Point 129°C to 130°C
Flash Point 24°C (75°F)
Refractive Index 1.452
Quantity 2500 g
UN Number UN2920
Beilstein 383568
Solubility Information Insoluble in water
Formula Weight 98.15
Percent Purity ≥98%
Chemical Name or Material Cyclohexene oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'