missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Selective inhibitor of eukaryotic (over prokaryotic) protein synthesis, blocking tRNA binding and release from ribosomes. Induces apoptosis in a variety of transformed and normal cell lines, including T cells. Competitively inhibits the PPIase hFKBP12 (Ki = 3.4 μM). Also inhibits ferroptosis. Antifungal antibiotic. Exhibits anti-MERS-CoV activity in Vero cells in vitro (IC50 = 0.16 μM).

Specifications
Specifications
CAS | 66-81-9 |
Molecular Formula | C15H23NO4 |
MDL Number | MFCD00082346 |
Quantity | 100 mg |
Synonym | cycloheximide, actidione, cycloheximide, naramycin a, kaken, actidion, actidone, hizarocin, naramycin, neocycloheximide |
InChI Key | YPHMISFOHDHNIV-QTEFRXOUNA-N |
SMILES | [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1 |
IUPAC Name | 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione |
Molecular Weight (g/mol) | 281.35 |
PubChem CID | 6197 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cycloheximide, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction