missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl phenyl ketone, 98%, Thermo Scientific Chemicals

28.64 € - 224.12 €

Chemical Identifiers

CAS 712-50-5
Molecular Formula C13H16O
Molecular Weight (g/mol) 188.27
MDL Number MFCD00001467
InChI Key BMFYCFSWWDXEPB-UHFFFAOYSA-N
Synonym cyclohexyl phenyl ketone, cyclohexyl phenyl methanone, benzoylcyclohexane, cyclohexylphenylketone, benzoyl cyclohexane, phenyl cyclohexyl ketone, methanone, cyclohexylphenyl, cyclohexyl-phenyl-methanone, usaf kf-3, cyclohexylphenyl ketone
PubChem CID 12837
IUPAC Name cyclohexyl(phenyl)methanone
SMILES C1CCC(CC1)C(=O)C2=CC=CC=C2
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11466576
View Documents
Thermo Scientific Alfa Aesar
B24816.06
5 g
28.64 €
5g
Please to purchase this item. Need a web account? Register with us today!
11476576
View Documents
Thermo Scientific Alfa Aesar
B24816.14
25 g
53.67 €
25g
Please to purchase this item. Need a web account? Register with us today!
11486576
View Documents
Thermo Scientific Alfa Aesar
B24816.22
100 g
224.12 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

712-50-5
188.27
BMFYCFSWWDXEPB-UHFFFAOYSA-N
12837
C1CCC(CC1)C(=O)C2=CC=CC=C2
C13H16O
MFCD00001467
cyclohexyl phenyl ketone, cyclohexyl phenyl methanone, benzoylcyclohexane, cyclohexylphenylketone, benzoyl cyclohexane, phenyl cyclohexyl ketone, methanone, cyclohexylphenyl, cyclohexyl-phenyl-methanone, usaf kf-3, cyclohexylphenyl ketone
cyclohexyl(phenyl)methanone

Specifications

712-50-5
165°C to 168°C (20 mmHg)
C13H16O
2046712
BMFYCFSWWDXEPB-UHFFFAOYSA-N
cyclohexyl(phenyl)methanone
12837
98%
52°C to 57°C
>110°C (230°F)
MFCD00001467
cyclohexyl phenyl ketone, cyclohexyl phenyl methanone, benzoylcyclohexane, cyclohexylphenylketone, benzoyl cyclohexane, phenyl cyclohexyl ketone, methanone, cyclohexylphenyl, cyclohexyl-phenyl-methanone, usaf kf-3, cyclohexylphenyl ketone
C1CCC(CC1)C(=O)C2=CC=CC=C2
188.27
188.27
Cyclohexyl phenyl ketone
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 211-923-5

missing translation for 'rtecsNumber' : OB1760000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.