missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 15461269
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87100-15-0
Molecular Formula C12H23BO2
Molecular Weight (g/mol) 210.124
MDL Number MFCD04038749
InChI Key OUEVCDGYTKLNMJ-UHFFFAOYSA-N
Synonym cyclohexylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-cyclohexyl-4,4,5,5-tetramethyl, cyclohexylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohexane, pubchem7924, cyclohexylboronic acid, pinacol eser, 1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohexane
PubChem CID 3668596
IUPAC Name 2-cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2CCCCC2

Specifications

Color Colorless
Quantity 25 g
Formula Weight 210.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cyclohexylboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'