missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylmagnesium chloride, 1.3M solution in THF/toluene, AcroSeal™

Product Code. 10381543 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10381543

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

CAS: 931-51-1 | C6H11ClMg | 142.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 931-51-1, 109-99-9, 108-88-3
Molecular Formula C6H11ClMg
Molecular Weight (g/mol) 142.91
MDL Number MFCD00003816
InChI Key WMJMABVHDMRMJA-UHFFFAOYSA-M
Synonym cyclohexylmagnesium chloride, magnesium, chlorocyclohexyl, magnesium,chlorocyclohexyl, cyclohexyl magnesium chloride, chloro cyclohexyl magnesium, chlorocyclohexylmagnesium, cyclohexylmagnesiumchloride, unii-oag686rdsu, cyclohexylmagnesium chloride solution, 2.0 m in diethyl ether, cyclohexylmagnesium cloride
PubChem CID 13605
IUPAC Name magnesium;cyclohexane;chloride
SMILES Cl[Mg]C1CCCCC1

Specifications

Color Brown
Density 0.9500g/mL
Flash Point −21°C
Packaging AcroSeal™ Glass Bottle
Linear Formula (C6H11)MgCl
Quantity 800 mL
Specific Gravity 0.95
Solubility Information Solubility in water: reacts violently with water
Formula Weight 142.91
Physical Form Solution
Chemical Name or Material Cyclohexylmagnesium chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexylmagnesium chloride, 1.3M solution in THF/toluene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.