missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylmethyl bromide, 96%, Thermo Scientific Chemicals

Product Code. 10253832
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2550-36-9
Molecular Formula C7H13Br
Molecular Weight (g/mol) 177.09
MDL Number MFCD00001509
InChI Key UUWSLBWDFJMSFP-UHFFFAOYSA-N
Synonym bromomethyl cyclohexane, cyclohexylmethyl bromide, cyclohexane, bromomethyl, bromocyclohexylmethane, cyclohexylmethylbromide, cyclohexylmethybromide, bromomethyl-cyclohexane, pubchem3103, 1-bromomethylcyclohexane
PubChem CID 137636
SMILES BrCC1CCCCC1

Specifications

Color Yellow
Density 1.2800g/mL
Boiling Point 76°C to 77°C (26.0 mmHg)
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula C6H11CH2Br
Refractive Index 1.4910 to 1.493
Quantity 5 g
Beilstein 05, IV, 100
Specific Gravity 1.28
IUPAC Name bromomethylcyclohexane
Formula Weight 177.08
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Cyclohexylmethyl bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'