Learn More
RETSCH Cyclone Mill Twister
Designed for processing foods and feeds for subsequent NIR analysis. Retsch™ Cyclone Mill Twister optimizes the reproducible sample preparation to NIR analysis, allowing for meaningful and reliable analysis results.
Brand:  RETSCH 20.831.0001
Product Code. 11730815
  • 9615.00 € / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Optimized form of rotor and grinding chamber generates an air jet which carries the ground sample through the integrated cyclone into the sample bottle
  • Air jet prevents the material from heating up, thus preserving the moisture content
  • Sieves guarantee optimum particle size distribution so that it is not necessary to recalibrate the NIR spectrometer
  • Rotor speed can be adjusted in 3 steps allowing for perfect adaptation to the sample requirements
  • Cleaning the mill is quick and easy as the air jet effects a complete discharge of the material from the grinding chamber
  • Ideal for grinding feeds, grains, forage and other similar products
  • 3 controlled speeds
  • Cyclone separator with 250mL collecting bottle for quick extraction of sample
  • No cross contamination thanks to easy cleaning
  • Convenient operating panel
  • Robust industrial motor 900w  with long lifetime
Specifications
Specifications
Cyclone Mill Twister
220/240 V, 50/60 Hz
For Grinding Prior to NIR Analysis
50/60 Hz
1-phase Power Connection
14 kg
283 x 449 x 427 mm
For Grinding Prior to NIR Analysis
427 mm
283 mm
220/240 V
449 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products