missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctanone, 98%, Thermo Scientific Chemicals

32.56 € - 271.35 €

Chemical Identifiers

CAS 502-49-8
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.2
MDL Number MFCD00001754
InChI Key IIRFCWANHMSDCG-UHFFFAOYSA-N
Synonym cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference
PubChem CID 10403
IUPAC Name cyclooctanone
SMILES C1CCCC(=O)CCC1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10350984
View Documents Promotion Details
Thermo Scientific Acros
111440250
25 g
32.56 €
25g
Please to purchase this item. Need a web account? Register with us today!
10385690
View Documents Promotion Details
Thermo Scientific
111441000
100 g
73.57 €
100g
Please to purchase this item. Need a web account? Register with us today!
10289393
View Documents Promotion Details
Thermo Scientific Acros
111445000
500 g
271.35 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

502-49-8
126.2
IIRFCWANHMSDCG-UHFFFAOYSA-N
10403
C1CCCC(=O)CCC1
C8H14O
MFCD00001754
cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference
cyclooctanone

Specifications

502-49-8
Colorless to White
195.0°C to 197.0°C
Authentic
Glass bottle
C8H14(=O)
07, 21
cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference
IIRFCWANHMSDCG-UHFFFAOYSA-N
cyclooctanone
10403
98%
Cyclooctanone, 98%
39.0°C to 44.0°C
0.9500g/mL
74°C
98%
C8H14O
MFCD00001754
0.95
Solubility in water: 15g/l (20°C). Other solubilities: soluble in alcohol,acetone and benzene
C1CCCC(=O)CCC1
126.2
126.2
Crystalline Low Melting Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 207-940-2

missing translation for 'rtecsNumber' : GX9800000

missing translation for 'tsca' : TSCA

Product Identifier
  • Cyclooctanone

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Certificates

RUO – Research Use Only