Learn More
Cyclooctanone, 98%, Thermo Scientific Chemicals
31.96 € - 305.52 €
Chemical Identifiers
CAS | 502-49-8 |
---|---|
Molecular Formula | C8H14O |
Molecular Weight (g/mol) | 126.199 |
MDL Number | MFCD00001754 |
InChI Key | IIRFCWANHMSDCG-UHFFFAOYSA-N |
Synonym | cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference |
PubChem CID | 10403 |
IUPAC Name | cyclooctanone |
SMILES | C1CCCC(=O)CCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11469946
|
Thermo Scientific Alfa Aesar
B20911.14 |
25 g |
31.96 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11479946
|
Thermo Scientific Alfa Aesar
B20911.22 |
100 g |
85.32 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489946
|
Thermo Scientific Alfa Aesar
B20911.36 |
500 g |
305.52 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclooctanone is a aliphatic cycle that exhibited inhibitory activity towards aldosterone synthase, a promising therapeutic target for the treatment of cardiovascular diseases related to abnormally high aldosterone level. It is also used in the synthesis of 14-membered lactones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water at 20°C 15g/L. Soluble in acetone, alcohol, chloroform, methanol and benzene.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
Chemical Identifiers
502-49-8 | |
126.199 | |
IIRFCWANHMSDCG-UHFFFAOYSA-N | |
10403 | |
C1CCCC(=O)CCC1 |
C8H14O | |
MFCD00001754 | |
cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference | |
cyclooctanone |
Specifications
502-49-8 | |
0.96 | |
72°C (161°F) | |
C8H14O | |
UN1759 | |
cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference | |
IIRFCWANHMSDCG-UHFFFAOYSA-N | |
cyclooctanone | |
10403 | |
98% |
37°C to 44°C | |
195°C to 197°C | |
Pleasant | |
MFCD00001754 | |
1280738 | |
Soluble in water at 20°C 15g/L. Soluble in acetone,alcohol,chloroform,methanol and benzene. | |
C1CCCC(=O)CCC1 | |
126.199 | |
126.2 | |
Cyclooctanone |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
missing translation for 'einecsNumber' : 207-940-2
missing translation for 'rtecsNumber' : GX9800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.