missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctylamine, 97%, Thermo Scientific Chemicals

Product Code. 10775941
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5452-37-9
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
MDL Number MFCD00001748
InChI Key HSOHBWMXECKEKV-UHFFFAOYSA-N
Synonym cyclooctylamine, aminocyclooctane, cyclooctyl amine, acmc-1ap2c, hsohbwmxeckekv-uhfffaoysa, cyclooctylamine, purum gc
PubChem CID 2903
IUPAC Name cyclooctanamine
SMILES C1CCCC(CCC1)N

Specifications

Melting Point -48°C
Color Colorless to Yellow
Density 0.9200g/mL
Boiling Point 190°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C8H15NH2
Refractive Index 1.4800 to 1.482
Quantity 5 g
Beilstein 12, IV, 128
Specific Gravity 0.92
Solubility Information Solubility in water: 10.2g/L (20°C)
Viscosity 5 mPa.s (20°C)
Formula Weight 127.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cyclooctylamine
Show More Show Less
compliance-icons
Product Identifier
  • Cyclooctylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'