missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanol, 99%, Thermo Scientific Chemicals

28.14 € - 83.11 €

Chemical Identifiers

CAS 96-41-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00001363
InChI Key XCIXKGXIYUWCLL-UHFFFAOYSA-N
Synonym hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol
PubChem CID 7298
ChEBI CHEBI:16133
IUPAC Name cyclopentanol
SMILES C1CCC(C1)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10716521
View Documents Promotion Details
Thermo Scientific Acros
111521000
100 g
28.14 €
100g
Please to purchase this item. Need a web account? Register with us today!
10696482
View Documents Promotion Details
Thermo Scientific Acros
111525000
500 g
83.11 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

96-41-3
86.13
XCIXKGXIYUWCLL-UHFFFAOYSA-N
7298
cyclopentanol
C5H10O
MFCD00001363
hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol
CHEBI:16133
C1CCC(C1)O

Specifications

96-41-3
Colorless to Yellow
139°C to 140°C
Authentic
Glass bottle
C5H9OH
MFCD00001363
15, 2737
hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol
XCIXKGXIYUWCLL-UHFFFAOYSA-N
cyclopentanol
7298
86.13
Liquid
-19°C
0.9400g/mL
51°C
99%
C5H10O
1.4520 to 1.454
06, 5
0.94
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol
C1CCC(C1)O
86.13
CHEBI:16133
99%
Cyclopentanol, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 202-504-8

missing translation for 'tsca' : TSCA

Product Identifier
  • Cyclopentanol

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only