missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanol, 99%, Thermo Scientific Chemicals

31.56 € - 320.60 €

Chemical Identifiers

CAS 96-41-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00001363
InChI Key XCIXKGXIYUWCLL-UHFFFAOYSA-N
Synonym hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol
PubChem CID 7298
ChEBI CHEBI:16133
IUPAC Name cyclopentanol
SMILES C1CCC(C1)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11472298
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15249.AE
100 mL
31.56 €
100mL
Please to purchase this item. Need a web account? Register with us today!
11482298
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15249.AP
500 mL
91.96 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11492298
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15249.0F
2500 mL
320.60 €
2500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cyclopentanol is a versatile compound used as a building block, intermediate and solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentanol is a versatile compound used as a building block, intermediate and solvent.

Solubility
Slightly soluble in water. Also soluble in ether, alcohols and acetone.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

96-41-3
86.134
XCIXKGXIYUWCLL-UHFFFAOYSA-N
7298
cyclopentanol
C5H10O
MFCD00001363
hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol
CHEBI:16133
C1CCC(C1)O

Specifications

96-41-3
0.95
51°C (123°F)
C5H10O
MFCD00001363
1900556
hydroxycyclopentane, cyclopentyl alcohol, 1-cyclopentanol, unii-1l43q07tbu, dsstox_cid_13371, dsstox_rid_79066, dsstox_gsid_33371, cydopentanol, 1-hydroxycyclopentyl, 1-cyclopentyl alcohol
XCIXKGXIYUWCLL-UHFFFAOYSA-N
cyclopentanol
7298
86.13
Cyclopentanol
-19°C
139°C to 141°C
Unpleasant
1.453
UN2244
14,2742
Slightly soluble in water. Also soluble in ether,alcohols and acetone.
C1CCC(C1)O
86.134
CHEBI:16133
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P243-P261-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P362-P374-P380-P501c

H226-H302-H310-H319

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYCLOPENTANOL

missing translation for 'einecsNumber' : 202-504-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Cyclopentanol

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : Cyclopentanol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only